Tisk…

3,4-Difluoro-N-methyl-benzylamine 97%

Dodavatel: Apollo Scientific

PC530055-1G PC530055-500MG
APOSPC530055-1GEA 12470 CZK
APOSPC530055-1G APOSPC530055-500MG
3,4-Difluoro-N-methyl-benzylamine 97%
3,4-Difluoro-N-methyl-benzylamine
Vzorec: C₈H₉F₂N
MW: 157,161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09043718
CAS číslo: 748124-46-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR