Tisk…

4-[2-(Trifluoromethyl)phenyl]-4-piperidinol hydrochloride 97%

Dodavatel: Apollo Scientific

PC530053-1G PC530053-500MG
APOSPC530053-1GEA 16170 CZK
APOSPC530053-1G APOSPC530053-500MG
4-[2-(Trifluoromethyl)phenyl]-4-piperidinol hydrochloride 97%
4-[2-(Trifluoromethyl)phenyl]-4-piperidinol hydrochloride
Vzorec: C₁₂H₁₄F₃NO·ClH
MW: 281,702 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28064040
CAS číslo: 69908-12-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR