4-(2,4-Difluorobenzyl)piperidine hydrochloride 97%
Dodavatel: Apollo Scientific
PC530052-1G
PC530052-250MG
PC530052-500MG
APOSPC530052-1GEA
22950
CZK
APOSPC530052-1G
APOSPC530052-250MG
APOSPC530052-500MG
4-(2,4-Difluorobenzyl)piperidine hydrochloride 97%
4-(2,4-Difluorobenzyl)piperidine hydrochloride
Vzorec:
C₁₂H₁₅F₂N·ClH MW: 247,712 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28144979 CAS číslo: 203860-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...