(R)-b-Amino-4-fluorobenzenepropanol 97%
Dodavatel: Apollo Scientific
PC530049-1G
PC530049-250MG
PC530049-500MG
APOSPC530049-1GEA
24800
CZK
APOSPC530049-1G
APOSPC530049-250MG
APOSPC530049-500MG
(R)-b-Amino-4-fluorobenzenepropanol 97%
(R)-b-Amino-4-fluorobenzenepropanol
Vzorec:
C₉H₁₂FNO MW: 169,196 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17214640 CAS číslo: 348619-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...