Tisk…

b-Amino-3,4-difluorobenzenepropanol 97%

Dodavatel: Apollo Scientific

PC530048-1G PC530048-250MG PC530048-500MG
APOSPC530048-1GEA 22320 CZK
APOSPC530048-1G APOSPC530048-250MG APOSPC530048-500MG
b-Amino-3,4-difluorobenzenepropanol 97%
b-Amino-3,4-difluorobenzenepropanol
Vzorec: C₉H₁₁F₂NO
Storage Temperature: Ambient
MDL Number: MFCD17171139
CAS číslo: 218450-18-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR