b-Amino-4-chloro-3-fluorobenzeneethanol 97%
Dodavatel: Apollo Scientific
Synonyms:
2-Amino-2-(4-chloro-3-fluorophenyl)ethanol
PC530046-1G
PC530046-250MG
APOSPC530046-1GEA
19810
CZK
APOSPC530046-1G
APOSPC530046-250MG
b-Amino-4-chloro-3-fluorobenzeneethanol 97%
b-Amino-4-chloro-3-fluorobenzeneethanol
Vzorec:
C₈H₉ClFNO MW: 189,615 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16040267 CAS číslo: 1250241-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...