Tisk…

b-Amino-3-(trifluoromethyl)benzeneethanol 97%

Dodavatel: Apollo Scientific

PC530043-1G PC530043-250MG PC530043-500MG
APOSPC530043-1GEA 26780 CZK
APOSPC530043-1G APOSPC530043-250MG APOSPC530043-500MG
b-Amino-3-(trifluoromethyl)benzeneethanol 97%
b-Amino-3-(trifluoromethyl)benzeneethanol
Vzorec: C₉H₁₀F₃NO
MW: 205,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11213673
CAS číslo: 1094709-76-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR