Tisk…

2-Amino-1-(2,4-difluorophenyl)ethanol hydrochloride 97%

Dodavatel: Apollo Scientific

PC530042-1G PC530042-500MG
APOSPC530042-1GEA 19910 CZK
APOSPC530042-1G APOSPC530042-500MG
2-Amino-1-(2,4-difluorophenyl)ethanol hydrochloride 97%
2-Amino-1-(2,4-difluorophenyl)ethanol hydrochloride
Vzorec: C₈H₉F₂NO·ClH
MW: 209,621 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09831938
CAS číslo: 51337-06-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR