(R)-b-Amino-3-fluoro-benzeneethanol 97%
Dodavatel: Apollo Scientific
Synonyms:
(R)-2-Amino-2-(3-fluorophenyl)ethanol
PC530038-1G
PC530038-250MG
APOSPC530038-1GEA
13460
CZK
APOSPC530038-1G
APOSPC530038-250MG
(R)-b-Amino-3-fluoro-benzeneethanol 97%
(R)-b-Amino-3-fluoro-benzeneethanol
Vzorec:
C₈H₁₀FNO MW: 155,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09256371 CAS číslo: 1213968-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...