6-Fluoro-L-tryptophan 97%
Dodavatel: Apollo Scientific
PC530036-1G
PC530036-250MG
PC530036-500MG
APOSPC530036-1GEA
9030
CZK
APOSPC530036-1G
APOSPC530036-250MG
APOSPC530036-500MG
6-Fluoro-L-tryptophan 97%
6-Fluoro-L-tryptophan
Vzorec:
C₁₁H₁₁FN₂O₂ MW: 222,216 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00137731 CAS číslo: 19310-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...