7-Fluoro-DL-tryptophan 97%
Dodavatel: Apollo Scientific
PC530034-1G
PC530034-250MG
PC530034-500MG
APOSPC530034-1GEA
13390
CZK
APOSPC530034-1G
APOSPC530034-250MG
APOSPC530034-500MG
7-Fluoro-DL-tryptophan 97%
7-Fluoro-DL-tryptophan
Vzorec:
C₁₁H₁₁FN₂O₂ MW: 222,216 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09264339 CAS číslo: 53314-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...