7-Fluoro-D-tryptophan 97%
Dodavatel: Apollo Scientific
PC530033-1G
PC530033-250MG
PC530033-500MG
APOSPC530033-1GEA
27910
CZK
APOSPC530033-1G
APOSPC530033-250MG
APOSPC530033-500MG
7-Fluoro-D-tryptophan 97%
7-Fluoro-D-tryptophan
Vzorec:
C₁₁H₁₁FN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09264338 CAS číslo: 138514-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...