(S)-3-Amino-3-(4-fluorophenyl)propionic acid 97%
Dodavatel: Apollo Scientific
PC530010-1G
PC530010-5G
APOSPC530010-1GEA
1990
CZK
APOSPC530010-1G
APOSPC530010-5G
(S)-3-Amino-3-(4-fluorophenyl)propionic acid 97%
(S)-3-Amino-3-(4-fluorophenyl)propionic acid
Vzorec:
C₉H₁₀FNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD03840458 CAS číslo: 151911-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...