Tisk…

(R)-3-Amino-3-(4-fluorophenyl)propionic acid 97%

Dodavatel: Apollo Scientific

PC530009-1G PC530009-5G
APOSPC530009-1GEA 4900 CZK
APOSPC530009-1G APOSPC530009-5G
(R)-3-Amino-3-(4-fluorophenyl)propionic acid 97%
(R)-3-Amino-3-(4-fluorophenyl)propionic acid
Vzorec: C₉H₁₀FNO₂
Storage Temperature: Ambient
MDL Number: MFCD03840459
CAS číslo: 151911-23-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR