Tisk…

2,3-Difluoro-D-phenylalanine 97%

Dodavatel: Apollo Scientific

PC530001-1G PC530001-250MG PC530001-500MG
APOSPC530001-1GEA 25130 CZK
APOSPC530001-1G APOSPC530001-250MG APOSPC530001-500MG
2,3-Difluoro-D-phenylalanine 97%
2,3-Difluoro-D-phenylalanine
Vzorec: C₉H₉F₂NO₂
MW: 201.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371989
CAS číslo: 266360-59-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR