Tisk…

3,5-Dichloro-4-(trifluoromethyl)benzeneboronic acid 95%

Dodavatel: Apollo Scientific

PC52826-1G PC52826-250MG
APOSPC52826-1GEA 22320 CZK
APOSPC52826-1G APOSPC52826-250MG
3,5-Dichloro-4-(trifluoromethyl)benzeneboronic acid 95%
3,5-Dichloro-4-(trifluoromethyl)benzeneboronic acid
Vzorec: C₇H₄BCl₂F₃O₂
MW: 258,818 g/mol
Storage Temperature: Ambient
CAS číslo: 2304546-23-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR