4-Bromo-2-chloro-3-fluorobenzaldehyde 95%
Dodavatel: Apollo Scientific
PC52714-1G
PC52714-250MG
PC52714-5G
APOSPC52714-1GEA
15480
CZK
APOSPC52714-1G
APOSPC52714-250MG
APOSPC52714-5G
4-Bromo-2-chloro-3-fluorobenzaldehyde 95%
4-Bromo-2-chloro-3-fluorobenzaldehyde
Vzorec:
C₇H₃BrClFO MW: 237.454 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28733869 CAS číslo: 1780699-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...