Tisk…

4-(1,1,2,2,2-Pentafluoroethoxy)benzonitrile

Dodavatel: Apollo Scientific

PC52712-1G PC52712-250MG PC52712-5G
APOSPC52712-1GEA 8700 CZK
APOSPC52712-1G APOSPC52712-250MG APOSPC52712-5G
4-(1,1,2,2,2-Pentafluoroethoxy)benzonitrile
4-(1,1,2,2,2-Pentafluoroethoxy)benzonitrile
Vzorec: C₉H₄F₅NO
MW: 237,126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32647610
CAS číslo: 2414146-37-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR