Tisk…

3-Amino-2-hydroxybenzotrifluoride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Amino-6-(trifluoromethyl)phenol, 2-Hydroxy-3-(trifluoromethyl)aniline

PC5269-1G PC5269-250MG
APOSPC5269-1GEA 6950 CZK
APOSPC5269-1G APOSPC5269-250MG
3-Amino-2-hydroxybenzotrifluoride
3-Amino-2-hydroxybenzotrifluoride
Vzorec: C₇H₆F₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD08532490
CAS číslo: 72534-45-3
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR