2-Hydroxy-5-(pentafluorothio)benzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
PC5251-100MG
PC5251-1G
PC5251-250MG
APOSPC5251-100MGEA
3310
CZK
APOSPC5251-100MG
APOSPC5251-1G
APOSPC5251-250MG
2-Hydroxy-5-(pentafluorothio)benzaldehyde
2-Hydroxy-5-(pentafluorothio)benzaldehyde
Vzorec:
C₇H₅F₅O₂S Storage Temperature: Ambient |
MDL Number:
MFCD11501046 CAS číslo: 1159512-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...