Tisk…

3-[3,5-Bis(trifluoromethyl)phenyl]propionic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 3-[3,5-Bis(trifluoromethyl)phenyl]propanoic acid

PC52474-1G PC52474-5G PC52474-25G
APOSPC52474-1GEA 3740 CZK
APOSPC52474-1G APOSPC52474-5G APOSPC52474-25G
3-[3,5-Bis(trifluoromethyl)phenyl]propionic acid 95%
3-[3,5-Bis(trifluoromethyl)phenyl]propionic acid
Vzorec: C₁₁H₈F₆O₂
MW: 286,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00674033
CAS číslo: 181772-16-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR