Tisk…

3-Bromo-4-hydroxy-5-nitrophenylsulphur pentafluoride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromo-6-nitro-4-(pentafluorothio)phenol, 3-Bromo-2-hydroxy-5-(pentafluorosulphanyl)nitrobenzene

PC5247-1G PC5247-250MG
APOSPC5247-1GEA 27710 CZK
APOSPC5247-1G APOSPC5247-250MG
3-Bromo-4-hydroxy-5-nitrophenylsulphur pentafluoride
3-Bromo-4-hydroxy-5-nitrophenylsulphur pentafluoride
Vzorec: C₆H₃BrF₅NO₃S
MW: 344.054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11501044
CAS číslo: 1159512-28-3
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR