6-Chloro-2-fluoro-3-methylcinnamic acid 95%
Dodavatel: Apollo Scientific
PC52469-1G
PC52469-250MG
PC52469-5G
APOSPC52469-1GEA
14150
CZK
APOSPC52469-1G
APOSPC52469-250MG
APOSPC52469-5G
6-Chloro-2-fluoro-3-methylcinnamic acid 95%
6-Chloro-2-fluoro-3-methylcinnamic acid
Vzorec:
C₁₀H₈ClFO₂ MW: 214,621 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03002788 CAS číslo: 682804-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...