4-Fluoro-2-methylcinnamic acid 95%
Dodavatel: Apollo Scientific
PC52468-1G
PC52468-250MG
PC52468-5G
PC52468-25G
APOSPC52468-1GEA
8700
CZK
APOSPC52468-1G
APOSPC52468-250MG
APOSPC52468-5G
APOSPC52468-25G
4-Fluoro-2-methylcinnamic acid 95%
4-Fluoro-2-methylcinnamic acid
Vzorec:
C₁₀H₉FO₂ MW: 180,176 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06205344 CAS číslo: 773129-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...