Tisk…

2,6-Dibromo-1-(pentafluorosulphur)benzene 95%

Dodavatel: Apollo Scientific

PC52458-1G PC52458-250MG PC52458-5G
APOSPC52458-1GEA 5390 CZK
APOSPC52458-1G APOSPC52458-250MG APOSPC52458-5G
2,6-Dibromo-1-(pentafluorosulphur)benzene 95%
2,6-Dibromo-1-(pentafluorosulphur)benzene
Vzorec: C₆H₃Br₂F₅S
MW: 361,953 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32708477
CAS číslo: 2514942-08-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR