4-(1,1,2,2,2-Pentafluoroethoxy)benzoic acid
Dodavatel: Apollo Scientific
PC52238-1G
PC52238-250MG
PC52238-5G
APOSPC52238-1GEA
8700
CZK
APOSPC52238-1G
APOSPC52238-250MG
APOSPC52238-5G
4-(1,1,2,2,2-Pentafluoroethoxy)benzoic acid
4-(1,1,2,2,2-Pentafluoroethoxy)benzoic acid
Vzorec:
C₉H₅F₅O₃ MW: 256,126 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11858215 CAS číslo: 79865-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...