Tisk…

6-Bromo-2-fluoro-3-iodopyridine 95%

Dodavatel: Apollo Scientific

PC52206-1G PC52206-250MG PC52206-5G
APOSPC52206-1GEA 11180 CZK
APOSPC52206-1G APOSPC52206-250MG APOSPC52206-5G
6-Bromo-2-fluoro-3-iodopyridine 95%
6-Bromo-2-fluoro-3-iodopyridine
Vzorec: C₅H₂BrFIN
MW: 301,883 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28139682
CAS číslo: 1599474-54-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR