2-Cyano-3,6-difluorpyridine 95%
Dodavatel: Apollo Scientific
PC52202-1G
PC52202-250MG
APOSPC52202-1GEA
22520
CZK
APOSPC52202-1G
APOSPC52202-250MG
2-Cyano-3,6-difluorpyridine 95%
2-Cyano-3,6-difluorpyridine
Vzorec:
C₆H₂F₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD13185483 CAS číslo: 1214332-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...