Ethyl 2-bromo-4,4-difluoro-3-oxobutanoate, Technical Grade
Dodavatel: Apollo Scientific
|
Varování
|
PC52197-1G
PC52197-250MG
APOSPC52197-1GEA
12200
CZK
APOSPC52197-1G
APOSPC52197-250MG
Ethyl 2-bromo-4,4-difluoro-3-oxobutanoate, Technical Grade
Ethyl 2-bromo-4,4-difluoro-3-oxobutanoate
Vzorec:
C₆H₇BrF₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD27940538 CAS číslo: 660840-16-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...