4-Iodo-3-methoxybenzotrifluoride 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Iodo-2-methoxy-4-(trifluoromethyl)benzene, 2-iodo-5-(trifluoromethyl)anisole
PC52196-1G
PC52196-250MG
PC52196-5G
APOSPC52196-1GEA
1660
CZK
APOSPC52196-1G
APOSPC52196-250MG
APOSPC52196-5G
4-Iodo-3-methoxybenzotrifluoride 95%
4-Iodo-3-methoxybenzotrifluoride
Vzorec:
C₈H₆F₃IO MW: 302,032 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18398114 CAS číslo: 1261752-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...