3-(1,1,1,3,3,3-Hexafluoro-2-hydroxypropan-2-yl)phenol 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC52191-1G
PC52191-250MG
APOSPC52191-1GEA
17360
CZK
APOSPC52191-1G
APOSPC52191-250MG
3-(1,1,1,3,3,3-Hexafluoro-2-hydroxypropan-2-yl)phenol 95%
3-(1,1,1,3,3,3-Hexafluoro-2-hydroxypropan-2-yl)phenol
Vzorec:
C₉H₆F₆O₂ Storage Temperature: Ambient |
MDL Number:
MFCD31812120 CAS číslo: 2366994-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...