Tisk…

2-Bromo-3-iodobenzal fluoride

Dodavatel: Apollo Scientific
Varování

PC52188-1G PC52188-250MG PC52188-5G
APOSPC52188-1GEA 14880 CZK
APOSPC52188-1G APOSPC52188-250MG APOSPC52188-5G
2-Bromo-3-iodobenzal fluoride
2-Bromo-3-iodobenzal fluoride
Vzorec: C₇H₄BrF₂I
MW: 332,912 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391105
CAS číslo: 1261552-67-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR