Tisk…

2-Bromo-6-chlorobenzal fluoride

Dodavatel: Apollo Scientific
Varování

PC52185-1G PC52185-5G
APOSPC52185-1GEA 5860 CZK
APOSPC52185-1G APOSPC52185-5G
2-Bromo-6-chlorobenzal fluoride
2-Bromo-6-chlorobenzal fluoride
Vzorec: C₇H₄BrClF₂
MW: 241.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698635
CAS číslo: 1214348-94-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR