5-Bromo-2,4-difluorobenzal fluoride
Dodavatel: Apollo Scientific
|
Varování
|
PC52179-1G
PC52179-250MG
APOSPC52179-1GEA
14880
CZK
APOSPC52179-1G
APOSPC52179-250MG
5-Bromo-2,4-difluorobenzal fluoride
5-Bromo-2,4-difluorobenzal fluoride
Vzorec:
C₇H₃BrF₄ MW: 242,996 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28737893 CAS číslo: 1804408-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...