5-Bromo-3-fluoro-2-hydrazinopyridine
Dodavatel: Apollo Scientific
|
Varování
|
PC52173-1G
PC52173-250MG
PC52173-5G
APOSPC52173-1GEA
8200
CZK
APOSPC52173-1G
APOSPC52173-250MG
APOSPC52173-5G
5-Bromo-3-fluoro-2-hydrazinopyridine
5-Bromo-3-fluoro-2-hydrazinopyridine
Vzorec:
C₅H₅BrFN₃ MW: 206,016 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09999913 CAS číslo: 1289084-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...