Tisk…

2,4-Difluoro-5-iodophenol

Dodavatel: Apollo Scientific
Varování

PC52128-1G PC52128-5G
APOSPC52128-1GEA 7940 CZK
APOSPC52128-1G APOSPC52128-5G
2,4-Difluoro-5-iodophenol
2,4-Difluoro-5-iodophenol
Vzorec: C₆H₃F₂IO
Storage Temperature: Ambient
MDL Number: MFCD28986198
CAS číslo: 1935521-44-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR