4-(Bromomethyl)-2-fluorobenzenesulphonamide 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC52125-1G
PC52125-250MG
PC52125-5G
PC52125-25G
APOSPC52125-1GEA
2200
CZK
APOSPC52125-1G
APOSPC52125-250MG
APOSPC52125-5G
APOSPC52125-25G
4-(Bromomethyl)-2-fluorobenzenesulphonamide 95%
4-(Bromomethyl)-2-fluorobenzenesulphonamide
Vzorec:
C₇H₇BrFNO₂S MW: 268,103 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28986199 CAS číslo: 1645275-47-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...