Tisk…

6-Amino-3-bromo-2-fluorobenzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC52104-1G PC52104-5G PC52104-25G
APOSPC52104-1GEA 910 CZK
APOSPC52104-1G APOSPC52104-5G APOSPC52104-25G
6-Amino-3-bromo-2-fluorobenzoic acid 95%
6-Amino-3-bromo-2-fluorobenzoic acid
Vzorec: C₇H₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD20482512
CAS číslo: 1036756-03-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR