Tisk…

4-(4-Fluorobenzyl)piperidine hydrochloride

Dodavatel: Apollo Scientific
Varování

PC52097-1G PC52097-25G PC52097-5G
APOSPC52097-1GEA 3940 CZK
APOSPC52097-1G APOSPC52097-25G APOSPC52097-5G
4-(4-Fluorobenzyl)piperidine hydrochloride
4-(4-Fluorobenzyl)piperidine hydrochloride
Vzorec: C₁₂H₁₆FN·ClH
MW: 229,723 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03840140
CAS číslo: 92822-03-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR