Tisk…

2-Chloro-5-(trifluoromethylthio)pyridine 97%

Dodavatel: Apollo Scientific

PC520896-1G PC520896-500MG
APOSPC520896-1GEA 31410 CZK
APOSPC520896-1G APOSPC520896-500MG
2-Chloro-5-(trifluoromethylthio)pyridine 97%
2-Chloro-5-(trifluoromethylthio)pyridine
Vzorec: C₆H₃ClF₃NS
Storage Temperature: Ambient
MDL Number: MFCD23382170
CAS číslo: 1204234-95-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR