4-Bromo-2-fluoro-6-iodobenzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
PC52085-1G
PC52085-250MG
APOSPC52085-1GEA
12400
CZK
APOSPC52085-1G
APOSPC52085-250MG
4-Bromo-2-fluoro-6-iodobenzonitrile
4-Bromo-2-fluoro-6-iodobenzonitrile
Vzorec:
C₇H₂BrFIN Storage Temperature: Ambient |
MDL Number:
MFCD28127023 CAS číslo: 796968-89-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...