Tisk…

3-Chloro-2-fluorobenzyl chloride

Dodavatel: Apollo Scientific

PC52084-1G PC52084-5G
APOSPC52084-1GEA 2450 CZK
APOSPC52084-1G APOSPC52084-5G
3-Chloro-2-fluorobenzyl chloride
3-Chloro-2-fluorobenzyl chloride
Vzorec: C₇H₅Cl₂F
Storage Temperature: Ambient
MDL Number: MFCD09744684
CAS číslo: 876384-47-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR