Tisk…

(E)-3-Butoxy-1-(4-(trifluoromethyl)phenyl)prop-2-en-1-one

Dodavatel: Apollo Scientific

PC520825-1G PC520825-250MG PC520825-500MG
APOSPC520825-1GEA 19210 CZK
APOSPC520825-1G APOSPC520825-250MG APOSPC520825-500MG
(E)-3-Butoxy-1-(4-(trifluoromethyl)phenyl)prop-2-en-1-one
(E)-3-Butoxy-1-(4-(trifluoromethyl)phenyl)prop-2-en-1-one
Vzorec: C₁₄H₁₅F₃O₂
MW: 272.263 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22683246
CAS číslo: 1375008-07-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR