Tisk…

(E)-3-Butoxy-1-(3-fluorophenyl)prop-2-en-1-one 95%

Dodavatel: Apollo Scientific

PC520824-1G PC520824-250MG PC520824-500MG
APOSPC520824-1GEA 14250 CZK
APOSPC520824-1G APOSPC520824-250MG APOSPC520824-500MG
(E)-3-Butoxy-1-(3-fluorophenyl)prop-2-en-1-one 95%
(E)-3-Butoxy-1-(3-fluorophenyl)prop-2-en-1-one
Vzorec: C₁₃H₁₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD22683241
CAS číslo: 1404196-17-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR