2-(Trifluoromethyl)cyclohexanecarboxylic acid
Dodavatel: Apollo Scientific
PC520811-1G
APOSPC520811-1GEA
25630
CZK
APOSPC520811-1G
2-(Trifluoromethyl)cyclohexanecarboxylic acid
2-(Trifluoromethyl)cyclohexanecarboxylic acid
Vzorec:
C₈H₁₁F₃O₂ MW: 196,167 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09811959 CAS číslo: 384-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...