4-[3-(4-Bromo-2-fluoro-phenoxy)propyl]morpholine ≥95%
Dodavatel: Apollo Scientific
PC520778-1G
PC520778-5G
APOSPC520778-1GEA
5000
CZK
APOSPC520778-1G
APOSPC520778-5G
4-[3-(4-Bromo-2-fluoro-phenoxy)propyl]morpholine ≥95%
4-[3-(4-Bromo-2-fluoro-phenoxy)propyl]morpholine
Vzorec:
C₁₃H₁₇BrFNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD22989488 CAS číslo: 944279-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...