Tisk…

2-Fluoro-1-(trifluoromethoxy)-3-(trifluoromethyl)benzene 95%

Dodavatel: Apollo Scientific

PC520774-250MG PC520774-500MG
APOSPC520774-250MGEA 11940 CZK
APOSPC520774-250MG APOSPC520774-500MG
2-Fluoro-1-(trifluoromethoxy)-3-(trifluoromethyl)benzene 95%
2-Fluoro-1-(trifluoromethoxy)-3-(trifluoromethyl)benzene
Vzorec: C₈H₃F₇O
MW: 248,098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22580889
CAS číslo: 1417567-37-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR