Tisk…

Dimethyl (1H,1H,2H,2H-tridecafluorooct-1-yl)phosphonate

Dodavatel: Apollo Scientific
Varování

PC52077-1G PC52077-250MG
APOSPC52077-1GEA 9920 CZK
APOSPC52077-1G APOSPC52077-250MG
Dimethyl (1H,1H,2H,2H-tridecafluorooct-1-yl)phosphonate
Dimethyl (1H,1H,2H,2H-tridecafluorooct-1-yl)phosphonate
Vzorec: C₁₀H₁₀F₁₃O₃P
Bod varu: 55…60 °C (0,03 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD28014017
CAS číslo: 61726-44-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR