Tisk…

3-Methoxybenzenesulphonyl fluoride 95%

Dodavatel: Apollo Scientific

PC520757-1G PC520757-5G
APOSPC520757-1GEA 5330 CZK
APOSPC520757-1G APOSPC520757-5G
3-Methoxybenzenesulphonyl fluoride 95%
3-Methoxybenzenesulphonyl fluoride
Vzorec: C₇H₇FO₃S
MW: 190,192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21881376
CAS číslo: 882670-26-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR