Hexafluoro-2-(2-hydroxyphenyl)propan-2-ol 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC520743-1G
PC520743-5G
APOSPC520743-1GEA
7410
CZK
APOSPC520743-1G
APOSPC520743-5G
Hexafluoro-2-(2-hydroxyphenyl)propan-2-ol 97%
Hexafluoro-2-(2-hydroxyphenyl)propan-2-ol
Vzorec:
C₉H₆F₆O₂ Storage Temperature: Ambient |
MDL Number:
MFCD18641877 CAS číslo: 836-78-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...